Assay Details
| Gene Symbol: EFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039256.2 | NP_001034345.2 | 15 - 16 | 1822 | 103 | |
| XM_025153836.1 | XP_025009604.1 | 14 - 15 | 1685 | 103 | ||
| GenBank mRNA | AJ851470.1 | - | 15 - 16 | 1823 | 103 | |
More Information
Set Membership: |
|

EFL1
Genomic Map