Assay Details
| Gene Symbol: EFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039256.2 | NP_001034345.2 | 16 - 17 | 1957 | 93 | |
| XM_025153836.1 | XP_025009604.1 | 15 - 16 | 1820 | 93 | ||
| GenBank mRNA | AJ851470.1 | - | 16 - 17 | 1958 | 93 | |
More Information
Set Membership: |
|

EFL1
Genomic Map