Assay Details
| Gene Symbol: EFL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039256.2 | NP_001034345.2 | 19 - 20 | 3295 | 57 | |
| XM_025153836.1 | XP_025009604.1 | 18 - 19 | 3158 | 57 | ||
| GenBank mRNA | AJ851470.1 | - | 19 - 20 | 3296 | 57 | |
More Information
Set Membership: |
|

EFL1
Genomic Map