Assay Details
| Gene Symbol: ETV7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004934884.3 | XP_004934941.1 | 129 | 72 | ||
| XM_025143806.1 | XP_024999574.1 | 44 | 72 | |||
| GenBank mRNA | BX950624.2 | - | 50 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004934884.3, XM_025143806.1 transcripts.
Set Membership: |
|

ETV7
Genomic Map