Assay Details
Gene Symbol: ETV7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004934881.3 | XP_004934938.1 | 1611 | 85 | ||
XM_004934882.3 | XP_004934939.1 | 1609 | 85 | |||
XM_004934884.3 | XP_004934941.1 | 242 | 85 | |||
XM_025143803.1 | XP_024999571.1 | 1610 | 85 | |||
XM_025143806.1 | XP_024999574.1 | 157 | 85 | |||
GenBank mRNA | BX950624.2 | - | 163 | 85 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
