Assay Details
| Gene Symbol: SOX30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414564.6 | XP_414564.1 | 1 - 2 | 822 | 102 | |
| GenBank mRNA | CR390418.1 | - | 1 - 2 | 486 | 102 | |
More Information
Set Membership: |
|

SOX30
Genomic Map