Assay Details
| Gene Symbol: TMEM150B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_421633.6 | XP_421633.3 | 5 - 6 | 4715 | 66 | |
| GenBank mRNA | CR390909.1 | - | 3 - 4 | 279 | 66 | |
More Information
Set Membership: |
|

TMEM150B
Genomic Map