Assay Details
| Gene Symbol: NINJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286843.2 | XP_015142329.2 | 2 - 3 | 344 | 65 | |
| GenBank mRNA | CR406066.1 | - | 2 - 3 | 322 | 65 | |
More Information
Set Membership: |
|

NINJ2
Genomic Map