Assay Details
| Gene Symbol: OSGIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004944148.3 | XP_004944205.1 | 6 - 7 | 936 | 75 | |
| XM_015279240.2 | XP_015134726.1 | 6 - 7 | 697 | 75 | ||
| XM_015279241.1 | XP_015134727.1 | 5 - 6 | 679 | 75 | ||
| XM_015279242.2 | XP_015134728.1 | 6 - 7 | 848 | 75 | ||
| XM_015279243.2 | XP_015134729.1 | 5 - 6 | 667 | 75 | ||
| XM_025154267.1 | XP_025010035.1 | 6 - 7 | 670 | 75 | ||
| XM_025154268.1 | XP_025010036.1 | 6 - 7 | 3104 | 75 | ||
| XM_414073.6 | XP_414073.2 | 5 - 6 | 625 | 75 | ||
| GenBank mRNA | CR523499.1 | - | 1 - 2 | 99 | 75 | |
More Information
Set Membership: |
|

OSGIN1
Genomic Map