Assay Details
| Gene Symbol: DCUN1D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277897.2 | XP_015133383.1 | 3 - 4 | 1082 | 100 | |
| XM_015277898.2 | XP_015133384.1 | 3 - 4 | 470 | 100 | ||
| XM_015277899.2 | XP_015133385.1 | 4 - 5 | 608 | 100 | ||
| XM_015277901.1 | XP_015133387.1 | 3 - 4 | 1084 | 100 | ||
| XM_025146715.1 | XP_025002483.1 | 3 - 4 | 656 | 100 | ||
| GenBank mRNA | BX932972.1 | - | 1 - 2 | 151 | 100 | |
| CR523518.1 | - | 3 - 4 | 422 | 100 | ||
More Information
Set Membership: |
|

DCUN1D2
Genomic Map