Assay Details
| Gene Symbol: VPS37B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415126.6 | XP_415126.1 | 3 - 4 | 422 | 90 | |
| GenBank mRNA | BX930631.1 | - | 3 - 4 | 374 | 90 | |
More Information
Set Membership: |
|

VPS37B
Genomic Map