Assay Details
| Gene Symbol: FAM159A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004936793.3 | XP_004936850.1 | 1 - 2 | 196 | 65 | |
| XM_422478.6 | XP_422478.1 | 1 - 2 | 205 | 65 | ||
| GenBank mRNA | BX931863.1 | - | 1 - 2 | 226 | 65 | |
More Information
Set Membership: |
|

FAM159A
Genomic Map