Assay Details
| Gene Symbol: FAM159A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422478.6 | XP_422478.1 | 2 - 3 | 354 | 149 | |
| GenBank mRNA | BX931863.1 | - | 2 - 3 | 375 | 149 | |
| BX932756.1 | - | 2 - 3 | 147 | |||
More Information
Set Membership: |
|

FAM159A
Genomic Map