Assay Details
| Gene Symbol: FAM159A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_422478.6 | XP_422478.1 | 3 - 4 | 434 | 136 | |
| GenBank mRNA | BX931863.1 | - | 3 - 4 | 455 | 136 | |
More Information
Set Membership: |
|

FAM159A
Genomic Map