Assay Details
| Gene Symbol: FAM20A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015279945.2 | XP_015135431.1 | 4 - 5 | 777 | 94 | |
| XM_015279946.2 | XP_015135432.1 | 4 - 5 | 785 | 94 | ||
| XM_015279947.2 | XP_015135433.1 | 3 - 4 | 728 | 94 | ||
| XM_015279948.2 | XP_015135434.1 | 4 - 5 | 782 | 94 | ||
| XM_015279949.2 | XP_015135435.1 | 4 - 5 | 781 | 94 | ||
| GenBank mRNA | BX934750.1 | - | 3 - 4 | 327 | 94 | |
| CR353950.1 | - | 1 - 2 | 57 | 94 | ||
More Information
Set Membership: |
|

FAM20A
Genomic Map