Assay Details
| Gene Symbol: HS3ST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294504.2 | XP_015149990.1 | 2 | 1569 | 100 | |
| GenBank mRNA | CR353015.1 | - | 1 | 1046 | 100 | |
More Information
Set Membership: |
|

HS3ST2
Genomic Map