Assay Details
| Gene Symbol: MCEE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293218.1 | NP_001280147.1 | 1 - 2 | 92 | 120 | |
| GenBank mRNA | BX930043.2 | - | 1 - 2 | 92 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001293218.1 transcripts.
Set Membership: |
|

MCEE
Genomic Map