Assay Details
Gene Symbol: CWC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277823.1 | NP_001264752.1 | 6 - 7 | 715 | 66 | |
NM_001277824.1 | NP_001264753.1 | 6 - 7 | 635 | 66 | ||
GenBank mRNA | AJ397998.1 | - | 2 - 3 | 210 | 66 | |
BU233359.1 | - | 2 - 3 | 137 | 66 | ||
BX932596.2 | - | 6 - 7 | 715 | 66 | ||
BX934467.2 | - | 6 - 7 | 608 | 66 | ||
BX936078.1 | - | 6 - 7 | 627 | 66 | ||
BX936226.2 | - | 6 - 7 | 636 | 66 | ||
CN219349.1 | - | 6 - 7 | 623 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
