Assay Details
| Gene Symbol: SNX20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293321.1 | NP_001280250.1 | 3 - 4 | 743 | 64 | |
| XM_025154160.1 | XP_025009928.1 | 2 - 3 | 329 | 64 | ||
| XM_025154161.1 | XP_025009929.1 | 3 - 4 | 442 | 64 | ||
| GenBank mRNA | CR353629.1 | - | 3 - 4 | 758 | 64 | |
More Information
Set Membership: |
|

SNX20
Genomic Map