Assay Details
| Gene Symbol: MPZL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252285.1 | NP_001239214.1 | 4 - 5 | 661 | 65 | |
| GenBank mRNA | CR353823.1 | - | 3 - 4 | 454 | 65 | |
More Information
Set Membership: |
|

MPZL3
Genomic Map