Assay Details
| Gene Symbol: ARHGDIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001277364.1 | NP_001264293.1 | 5 - 6 | 505 | 76 | |
| XM_025148365.1 | XP_025004133.1 | 5 - 6 | 657 | 76 | ||
| GenBank mRNA | AJ399417.1 | - | 1 - 2 | 70 | 76 | |
| AM064639.1 | - | 5 - 6 | 463 | 76 | ||
| AM068818.1 | - | 5 - 6 | 435 | 76 | ||
| BX929293.2 | - | 5 - 6 | 473 | 76 | ||
| BX929376.2 | - | 5 - 6 | 481 | 76 | ||
| CD763411.1 | - | 5 - 6 | 505 | 76 | ||
More Information
Set Membership: |
|

ARHGDIB
Genomic Map