Assay Details
| Gene Symbol: CYP8B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147312.1 | XP_025003080.1 | 1 - 2 | 75 | 86 | |
| GenBank mRNA | BX934337.2 | - | 1 - 2 | 51 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025147312.1 transcripts.
Set Membership: |
|

CYP8B1
Genomic Map