Assay Details
| Gene Symbol: CYP8B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147312.1 | XP_025003080.1 | 2 - 3 | 133 | 110 | |
| XM_418499.6 | XP_418499.2 | 2 - 3 | 262 | 110 | ||
| GenBank mRNA | BX934337.2 | - | 2 - 3 | 109 | 110 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025147312.1 transcripts.
Set Membership: |
|

CYP8B1
Genomic Map