Assay Details
| Gene Symbol: FAM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416736.5 | XP_416736.2 | 7 - 8 | 729 | 82 | |
| GenBank mRNA | BX935728.2 | - | 8 - 9 | 777 | 82 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FAM3B| Gene Symbol: FAM3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_416736.5 | XP_416736.2 | 7 - 8 | 729 | 82 | |
| GenBank mRNA | BX935728.2 | - | 8 - 9 | 777 | 82 | |
Set Membership: |
|