Assay Details
| Gene Symbol: TMEM38B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004949357.3 | XP_004949414.1 | 5 - 6 | 744 | 82 | |
| XM_015280517.2 | XP_015136003.1 | 6 - 7 | 935 | 82 | ||
| XM_015280518.2 | XP_015136004.1 | 5 - 6 | 742 | 82 | ||
| XM_424887.5 | XP_424887.1 | 4 - 5 | 560 | 82 | ||
| GenBank mRNA | BX950655.2 | - | 4 - 5 | 538 | 82 | |
More Information
Set Membership: |
|

TMEM38B
Genomic Map