Assay Details
| Gene Symbol: TAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145060.1 | XP_025000828.1 | 1 - 2 | 335 | 70 | |
| GenBank mRNA | CR387616.1 | - | 1 - 2 | 131 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025145060.1 transcripts.
Set Membership: |
|

TAL2
Genomic Map