Assay Details
| Gene Symbol: SNTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142847.1 | NP_001136319.1 | 1 - 2 | 130 | ||
| GenBank mRNA | BX933188.1 | - | 1 - 2 | 251 | 130 | |
More Information
Set Membership: |
|

SNTN
Genomic Map