Assay Details
| Gene Symbol: SNTN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142847.1 | NP_001136319.1 | 3 - 4 | 275 | 70 | |
| GenBank mRNA | BX933188.1 | - | 3 - 4 | 419 | 70 | |
| BX935672.1 | - | 2 - 3 | 566 | 70 | ||
More Information
Set Membership: |
|

SNTN
Genomic Map