Assay Details
| Gene Symbol: GABRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015285484.2 | XP_015140970.1 | 3 - 4 | 459 | 76 | |
| XM_025149993.1 | XP_025005761.1 | 3 - 4 | 456 | 76 | ||
| XM_025149995.1 | XP_025005763.1 | 3 - 4 | 459 | 76 | ||
| XM_420725.5 | XP_420725.2 | 3 - 4 | 431 | 76 | ||
| GenBank mRNA | Z25467.1 | - | 306 | 76 | ||
More Information
Set Membership: |
|

GABRG1
Genomic Map