Assay Details
| Gene Symbol: GLIS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291201.2 | XP_015146687.1 | 5 - 6 | 1875 | 87 | |
| GenBank mRNA | CR390886.1 | - | 2 - 3 | 503 | 87 | |
More Information
Set Membership: |
|

GLIS1
Genomic Map