Assay Details
| Gene Symbol: LIMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006330.2 | NP_001006330.2 | 2 - 3 | 200 | 70 | |
| XM_015299316.2 | XP_015154802.1 | 2 - 3 | 335 | 70 | ||
| XM_015299317.2 | XP_015154803.1 | 2 - 3 | 252 | 70 | ||
| XM_015299318.2 | XP_015154804.1 | 2 - 3 | 196 | 70 | ||
| XM_015299320.2 | XP_015154806.1 | 2 - 3 | 300 | 70 | ||
| XM_015299321.2 | XP_015154807.1 | 2 - 3 | 370 | 70 | ||
| XM_025143969.1 | XP_024999737.1 | 2 - 3 | 288 | 70 | ||
| GenBank mRNA | AJ453706.1 | - | 2 - 3 | 200 | 70 | |
| AJ721104.1 | - | 2 - 3 | 199 | 70 | ||
More Information
Set Membership: |
|

LIMD2
Genomic Map