Assay Details
| Gene Symbol: EIF3M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006406.1 | NP_001006406.1 | 1 - 2 | 98 | 72 | |
| GenBank mRNA | AJ720570.1 | - | 1 - 2 | 98 | 72 | |
| CR352624.1 | - | 1 - 2 | 82 | 72 | ||
More Information
Set Membership: |
|

EIF3M
Genomic Map