Assay Details
| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204553.4 | NP_989884.2 | 1 - 2 | 701 | 72 | |
| GenBank mRNA | AJ404652.1 | - | 1 - 2 | 550 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SFRP1| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204553.4 | NP_989884.2 | 1 - 2 | 701 | 72 | |
| GenBank mRNA | AJ404652.1 | - | 1 - 2 | 550 | 72 | |
Set Membership: |
|