Assay Details
| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204553.4 | NP_989884.2 | 2 - 3 | 773 | 91 | |
| GenBank mRNA | AJ404652.1 | - | 2 - 3 | 622 | 91 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SFRP1| Gene Symbol: SFRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204553.4 | NP_989884.2 | 2 - 3 | 773 | 91 | |
| GenBank mRNA | AJ404652.1 | - | 2 - 3 | 622 | 91 | |
Set Membership: |
|