Assay Details
Gene Symbol: SULT1C3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204601.2 | NP_989932.1 | 2 - 3 | 223 | 117 | |
XM_015277822.2 | XP_015133308.1 | 3 - 4 | 331 | 117 | ||
GenBank mRNA | AJ416889.1 | - | 2 - 3 | 224 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
