Assay Details
| Gene Symbol: SLC16A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204663.1 | NP_989994.1 | 2 - 3 | 326 | 71 | |
| GenBank mRNA | AF204396.2 | - | 2 - 3 | 326 | 71 | |
More Information
Set Membership: |
|

SLC16A3
Genomic Map