Assay Details
| Gene Symbol: NEGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204856.1 | NP_990187.1 | 3 - 4 | 532 | 71 | |
| XM_025152755.1 | XP_025008523.1 | 3 - 4 | 650 | 71 | ||
| GenBank mRNA | AJ132998.1 | - | 3 - 4 | 532 | 71 | |
| AJ132999.1 | - | 3 - 4 | 532 | 71 | ||
More Information
Set Membership: |
|

NEGR1
Genomic Map