Assay Details
| Gene Symbol: WNT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204887.1 | NP_990218.1 | 4 - 5 | 957 | 67 | |
| XM_015292954.2 | XP_015148440.1 | 4 - 5 | 1544 | 67 | ||
| GenBank mRNA | AB006014.1 | - | 4 - 5 | 957 | 67 | |
| KX897002.1 | - | 3 - 4 | 577 | 67 | ||
More Information
Set Membership: |
|

WNT5A
Genomic Map