Assay Details
| Gene Symbol: KCNA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204899.2 | NP_990230.2 | 2 | 2407 | 90 | |
| GenBank mRNA | AF084460.1 | - | 1 | 2156 | 90 | |
| U82365.1 | - | 1 | 2107 | 90 | ||
More Information
Set Membership: |
|

KCNA4
Genomic Map