Assay Details
| Gene Symbol: CALB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205513.1 | NP_990844.1 | 2 - 3 | 259 | 60 | |
| GenBank mRNA | EU404189.1 | - | 2 - 3 | 157 | 60 | |
| GU230771.1 | - | 1 - 2 | 38 | 60 | ||
| M14230.1 | - | 2 - 3 | 259 | 60 | ||
| X03343.1 | - | 257 | 60 | |||
More Information
Set Membership: |
|

CALB1
Genomic Map