Assay Details
| Gene Symbol: WNT8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205531.1 | NP_990862.1 | 5 - 6 | 602 | 61 | |
| GenBank mRNA | AB193181.1 | - | 5 - 6 | 514 | 61 | |
| U02097.1 | - | 5 - 6 | 602 | 61 | ||
More Information
Set Membership: |
|

WNT8A
Genomic Map