Assay Details
| Gene Symbol: DFNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006361.1 | NP_001006361.1 | 6 - 7 | 867 | 83 | |
| XM_015281445.2 | XP_015136931.1 | 5 - 6 | 808 | 83 | ||
| XM_015281446.2 | XP_015136932.1 | 6 - 7 | 919 | 83 | ||
| XM_015281447.2 | XP_015136933.1 | 5 - 6 | 1802 | 83 | ||
| XM_015281448.1 | XP_015136934.1 | 4 - 5 | 687 | 83 | ||
| GenBank mRNA | AJ720558.1 | - | 6 - 7 | 867 | 83 | |
More Information
Set Membership: |
|

DFNA5
Genomic Map