Assay Details
| Gene Symbol: TMEM106B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012558.1 | NP_001012576.1 | 6 - 7 | 739 | 101 | |
| XM_015281406.2 | XP_015136892.1 | 6 - 7 | 815 | 101 | ||
| GenBank mRNA | AJ851510.1 | - | 6 - 7 | 739 | 101 | |
More Information
Set Membership: |
|

TMEM106B
Genomic Map