Assay Details
| Gene Symbol: HES5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012695.1 | NP_001012713.1 | 1 - 2 | 99 | 63 | |
| GenBank mRNA | AY916777.1 | - | 1 - 2 | 99 | 63 | |
| BX929926.1 | - | 1 - 2 | 103 | 63 | ||
More Information
Set Membership: |
|

HES5
Genomic Map