Assay Details
| Gene Symbol: BSDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012842.1 | NP_001012860.1 | 5 - 6 | 433 | 57 | |
| XM_025142996.1 | XP_024998764.1 | 4 - 5 | 588 | 57 | ||
| GenBank mRNA | AJ720778.1 | - | 5 - 6 | 433 | 57 | |
More Information
Set Membership: |
|

BSDC1
Genomic Map