Assay Details
| Gene Symbol: DNAJC18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025609.1 | NP_001020780.1 | 5 - 6 | 669 | 70 | |
| GenBank mRNA | CR522888.1 | - | 5 - 6 | 669 | 70 | |
| CR523884.1 | - | 3 - 4 | 423 | 70 | ||
More Information
Set Membership: |
|

DNAJC18
Genomic Map