Assay Details
| Gene Symbol: TMEM104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031107.1 | NP_001026278.1 | 4 - 5 | 343 | 76 | |
| XM_015295395.2 | XP_015150881.1 | 4 - 5 | 346 | 76 | ||
| XM_015295396.2 | XP_015150882.1 | 4 - 5 | 364 | 76 | ||
| XM_015295397.2 | XP_015150883.1 | 4 - 5 | 450 | 76 | ||
| GenBank mRNA | AJ851664.1 | - | 4 - 5 | 343 | 76 | |
More Information
Set Membership: |
|

TMEM104
Genomic Map