Assay Details
| Gene Symbol: ABHD17B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031423.1 | NP_001026594.1 | 2 - 3 | 763 | 96 | |
| XM_015280288.2 | XP_015135774.1 | 2 - 3 | 758 | 96 | ||
| GenBank mRNA | AJ720633.1 | - | 2 - 3 | 763 | 96 | |
More Information
Set Membership: |
|

ABHD17B
Genomic Map