Assay Details
| Gene Symbol: FRZB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204772.2 | NP_990103.1 | 5 - 6 | 873 | 62 | |
| GenBank mRNA | AF218057.1 | - | 5 - 6 | 870 | 62 | |
| DQ017062.1 | - | 5 - 6 | 873 | 62 | ||
More Information
Set Membership: |
|

FRZB
Genomic Map