Assay Details
| Gene Symbol: EREG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001203.1 | NP_001001203.1 | 1 - 2 | 147 | 72 | |
| GenBank mRNA | AY582132.1 | - | 1 - 2 | 147 | 72 | |
| BX930141.1 | - | 1 - 2 | 105 | 72 | ||
| BX935342.2 | - | 1 - 2 | 149 | 72 | ||
More Information
Set Membership: |
|

EREG
Genomic Map